1-Boc-piperazine-2-carboxylic acid 97%

Synonyms: 1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid

BD36554-25G BD36554-1G BD36554-5G BD36554-10G BD36554-250MG
BLDPBD36554-25GEA 456 EUR
BLDPBD36554-25G BLDPBD36554-1G BLDPBD36554-5G BLDPBD36554-10G BLDPBD36554-250MG
1-Boc-piperazine-2-carboxylic acid 97%
1-Boc-piperazine-2-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₀H₁₈N₂O₄
MW: 230,2600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02682403
CAS Number: 1214196-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR