1-Bromo-3-fluoro-5-nitrobenzene 98%

BLDPBD37606-500GEA 1560 EUR
BLDPBD37606-500G BLDPBD37606-5G BLDPBD37606-10G BLDPBD37606-25G BLDPBD37606-1G BLDPBD37606-100G
1-Bromo-3-fluoro-5-nitrobenzene 98%
1-Bromo-3-fluoro-5-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₃BrFNO₂
MW: 220,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09955454
CAS Number: 7087-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR