Lansoprazole 98%
|
|
Synonyms:
2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
BLDPBD41116-5GEA
26.3
EUR
BLDPBD41116-5G
BLDPBD41116-25G
BLDPBD41116-1G
BLDPBD41116-100G
Lansoprazole 98%
Lansoprazole
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₆H₁₄F₃N₃O₂S MW: 369,36 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00866873 CAS Number: 103577-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...