(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: (S)-tert-Butyl 4-isopropyl-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

BLDPBD447678-100MGEA 117 EUR
BLDPBD447678-100MG BLDPBD447678-250MG BLDPBD447678-1G
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected 98%
(4S)-2,2-Dioxido-4- isopropyl -1,2,3-oxathiazolidine, N-BOC protected

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₉NO₅S
MW: 265,327 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17018789
CAS Number: 1206227-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR