5-Hydroxy-N-methylpyridine-2-carboxamide 97%

Synonyms: 5-Hydroxy-2-(methylcarbamoyl)pyridine5-Hydroxy-N-methylpicolinamide

BLDPBD448127-100MGEA 57.8 EUR
BLDPBD448127-100MG BLDPBD448127-250MG
5-Hydroxy-N-methylpyridine-2-carboxamide 97%
5-Hydroxy-N-methylpyridine-2-carboxamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₈N₂O₂
MW: 152,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977447
CAS Number: 859538-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR