(R)-3-Amino-4-methylpentanoic acid 95%

Supplier: BLD PHARMATECH GMBH

BLDPBD47823-1GEA 125 EUR
BLDPBD47823-1G BLDPBD47823-100MG BLDPBD47823-250MG
(R)-3-Amino-4-methylpentanoic acid 95%
(R)-3-Amino-4-methylpentanoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₃NO₂
MW: 131,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01076231
CAS Number: 75992-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR