Ethyl 7-bromo-1-benzothiophene-2-carboxylate 97%

Synonyms: Ethyl 7-bromobenzo[b]thiophene-2-carboxylate

BD483990-5G BD483990-250MG BD483990-1G
BLDPBD483990-5GEA 449 EUR
BLDPBD483990-5G BLDPBD483990-250MG BLDPBD483990-1G
Ethyl 7-bromo-1-benzothiophene-2-carboxylate 97%
Ethyl 7-bromo-1-benzothiophene-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₉BrO₂S
MW: 285,157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17168278
CAS Number: 1355171-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR