3-Fluoro-5-methylphenylboronic acid 98%

Synonyms: (3-Fluoro-5-methylphenyl)boronic acid3-Fluoro-5-methylbenzeneboronic acid

BLDPBD4897-250MGEA 17.9 EUR
BLDPBD4897-250MG BLDPBD4897-1G BLDPBD4897-5G
3-Fluoro-5-methylphenylboronic acid 98%
3-Fluoro-5-methylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₈BFO₂
MW: 153,95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363785
CAS Number: 850593-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR