(9,9-Diphenylfluoren-4-yl)boronic acid 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: (9,9-Diphenyl-9H-fluoren-4-yl)boronic acid

BLDPBD491058-250MGEA 7.4 EUR
BLDPBD491058-250MG BLDPBD491058-100MG BLDPBD491058-5G BLDPBD491058-1G
(9,9-Diphenylfluoren-4-yl)boronic acid 97%
(9,9-Diphenylfluoren-4-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₅H₁₉BO₂
MW: 362,228 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29077672
CAS Number: 1224976-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR