4-(1,3,4-Oxadiazol-2-yl)benzaldehyde 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-(4-Formylphenyl)-1,3,4-oxadiazole

BLDPBD493518-100MGEA 62 EUR
BLDPBD493518-100MG BLDPBD493518-1G BLDPBD493518-250MG
4-(1,3,4-Oxadiazol-2-yl)benzaldehyde 98%
4-(1,3,4-Oxadiazol-2-yl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆N₂O₂
MW: 174,156 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18909697
CAS Number: 545424-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR