2'-Bromo-5'-methoxyacetophenone 98%
|
|
Synonyms:
1-(2-Bromo-5-methoxyphenyl)ethanone
BLDPBD49853-1GEA
60.9
EUR
BLDPBD49853-1G
BLDPBD49853-5G
BLDPBD49853-250MG
BLDPBD49853-25G
2'-Bromo-5'-methoxyacetophenone 98%
2'-Bromo-5'-methoxyacetophenone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₉BrO₂ MW: 229,0706 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683853 CAS Number: 6342-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...