1-(4-Bromobenzyl)homopiperazine 97%

Synonyms: 1-(4-Bromobenzyl)-1,4-diazepane

BD50847-1G BD50847-250MG BD50847-100MG
BLDPBD50847-1GEA 414 EUR
BLDPBD50847-1G BLDPBD50847-250MG BLDPBD50847-100MG
1-(4-Bromobenzyl)homopiperazine 97%
1-(4-Bromobenzyl)homopiperazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₂H₁₇BrN₂
MW: 269,181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407498
CAS Number: 690632-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR