5-Carboxyphthalide 98%
|
|
Synonyms:
1-Oxo-1,3-dihydroisobenzofuran-5-carboxylic acid
BLDPBD54065-5GEA
298
EUR
BLDPBD54065-5G
BLDPBD54065-25G
BLDPBD54065-1G
BLDPBD54065-250MG
5-Carboxyphthalide 98%
5-Carboxyphthalide
Storage condition: Sealed in dry, 2-8°C
Formula:
C₉H₆O₄ MW: 178,1400 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04117971 CAS Number: 4792-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...