5-Amino-1,3-dihydrobenzo[c]furan 97%

Synonyms: 1,3-Dihydroisobenzofuran-5-amine,5-Amino-1,3-dihydro-2-benzofuran,1,3-Dihydro-2-benzofuran-5-amine

BD54082-1G BD54082-250MG BD54082-5G BD54082-25G BD54082-10G
BLDPBD54082-1GEA 88.2 EUR
BLDPBD54082-1G BLDPBD54082-250MG BLDPBD54082-5G BLDPBD54082-25G BLDPBD54082-10G
5-Amino-1,3-dihydrobenzo[c]furan 97%
5-Amino-1,3-dihydrobenzo[c]furan

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₉NO
MW: 135,163 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864561
CAS Number: 61964-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR