1,4-Thioxane 98%

Synonyms: 1,4-Oxathiane

BLDPBD54810-25GEA 49.4 EUR
BLDPBD54810-25G BLDPBD54810-5G
1,4-Thioxane 98%
1,4-Thioxane

Storage condition: Sealed in dry, Room Temperature

Formula: C₄H₈OS
MW: 104,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006573
CAS Number: 15980-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR