5-Cyano-2-fluorobenzaldehyde 95%

Synonyms: 4-Fluoro-3-formylbenzonitrile

BD5661-250MG BD5661-1G BD5661-5G BD5661-10G BD5661-100G BD5661-25G
BLDPBD5661-250MGEA 7.4 EUR
BLDPBD5661-250MG BLDPBD5661-1G BLDPBD5661-5G BLDPBD5661-10G BLDPBD5661-100G BLDPBD5661-25G
5-Cyano-2-fluorobenzaldehyde 95%
5-Cyano-2-fluorobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₄FNO
MW: 149,1219 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974126
CAS Number: 146137-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR