1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%

Supplier: BLD PHARMATECH GMBH

BLDPBD566284-100MGEA 56.7 EUR
BLDPBD566284-100MG BLDPBD566284-250MG BLDPBD566284-1G
1-(3-Bromophenyl)cyclopentanecarboxylic acid 97%
1-(3-Bromophenyl)cyclopentanecarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₃BrO₂
MW: 269,134 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00573784
CAS Number: 143328-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR