3'-Bromo-4'-iodoacetophenone 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1-(3-Bromo-4-iodophenyl)ethanone

BLDPBD567756-1GEA 58.8 EUR
BLDPBD567756-1G BLDPBD567756-250MG
3'-Bromo-4'-iodoacetophenone 97%
3'-Bromo-4'-iodoacetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₆BrIO
MW: 324,941 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391610
CAS Number: 945907-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR