Dodecane-1,12-diamine, N-BOC protected 96%

Synonyms: tert-Butyl (12-aminododecyl)carbamate,tert-Butyl (12-aminododec-1-yl)carbamate,1,12-Diaminododecane, N-BOC protected

BD56874-100MG BD56874-250MG BD56874-1G BD56874-5G
BLDPBD56874-100MGEA 75.6 EUR
BLDPBD56874-100MG BLDPBD56874-250MG BLDPBD56874-1G BLDPBD56874-5G
Dodecane-1,12-diamine, N-BOC protected 96%
Dodecane-1,12-diamine, N-BOC protected

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₇H₃₆N₂O₂
MW: 300,48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02094503
CAS Number: 109792-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR