1-Chloro-6-methylisoquinoline 97%

BLDPBD57878-250MGEA 116 EUR
BLDPBD57878-250MG BLDPBD57878-1G BLDPBD57878-100MG
1-Chloro-6-methylisoquinoline 97%
1-Chloro-6-methylisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₈ClN
MW: 177,6302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110400
CAS Number: 209286-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR