4-Methoxy-1H-benzimidazole 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 7-Methoxy-1H-benzo[d]imidazole

BLDPBD58606-1GEA 89.3 EUR
BLDPBD58606-1G BLDPBD58606-250MG BLDPBD58606-5G
4-Methoxy-1H-benzimidazole 98%
4-Methoxy-1H-benzimidazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈N₂O
MW: 148,162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11973699
CAS Number: 27080-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR