Sodium-1-hexadecanesulphonate 98%

Synonyms: Sodium hexadecane-1-sulphonateHexadecane-1-sulphonic acid sodium salt

BLDPBD58702-25GEA 13.7 EUR
BLDPBD58702-25G BLDPBD58702-500G BLDPBD58702-100G
Sodium-1-hexadecanesulphonate 98%
Sodium 1-hexadecanesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₆H₃₃NaSO₃
MW: 328,49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007528
CAS Number: 15015-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR