1H-Imidazo[4,5-b]pyrazine 97%

BLDPBD58734-100MGEA 39.9 EUR
BLDPBD58734-100MG BLDPBD58734-250MG
1H-Imidazo[4,5-b]pyrazine 97%
1H-Imidazo[4,5-b]pyrazine

Storage condition: Sealed in dry, Room Temperature

Formula: C₅H₄N₄
MW: 120,112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669465
CAS Number: 273-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR