3-Chloro-4-methylbenzonitrile 97%

BD5882-10G BD5882-5G BD5882-25G BD5882-1G
BLDPBD5882-10GEA 27.3 EUR
BLDPBD5882-10G BLDPBD5882-5G BLDPBD5882-25G BLDPBD5882-1G
3-Chloro-4-methylbenzonitrile 97%
3-Chloro-4-methylbenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₆ClN
MW: 151,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001799
CAS Number: 21423-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR