5-Bromo-2-phenyl-1H-indole 97%
|
|
BLDPBD58839-5GEA
369
EUR
BLDPBD58839-5G
BLDPBD58839-250MG
BLDPBD58839-1G
BLDPBD58839-100MG
5-Bromo-2-phenyl-1H-indole 97%
5-Bromo-2-phenyl-1H-indole
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₄H₁₀BrN MW: 272,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04151729 CAS Number: 83515-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...