4-Methoxy-2-nitrobenzaldehyde 98%

BD6233-250MG BD6233-100MG BD6233-25G BD6233-100G BD6233-5G BD6233-10G BD6233-1G
BLDPBD6233-250MGEA 8.4 EUR
BLDPBD6233-250MG BLDPBD6233-100MG BLDPBD6233-25G BLDPBD6233-100G BLDPBD6233-5G BLDPBD6233-10G BLDPBD6233-1G
4-Methoxy-2-nitrobenzaldehyde 98%
4-Methoxy-2-nitrobenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₇NO₄
MW: 181,1455 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024254
CAS Number: 22996-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR