2-Benzyl-3-hydroxypropanenitrile 97%

BLDPBD625896-250MGEA 111 EUR
BLDPBD625896-250MG BLDPBD625896-5G BLDPBD625896-1G BLDPBD625896-100MG
2-Benzyl-3-hydroxypropanenitrile 97%
2-Benzyl-3-hydroxypropanenitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₁NO
MW: 161,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18294353
CAS Number: 2601-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR