3-Pyridineacetamide 96%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-(Pyridin-3-yl)acetamide

BLDPBD63501-25GEA 386 EUR
BLDPBD63501-25G BLDPBD63501-1G BLDPBD63501-250MG BLDPBD63501-5G
3-Pyridineacetamide 96%
3-Pyridineacetamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₈N₂O
MW: 136,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236789
CAS Number: 3724-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR