2-Ethynyl-5-methoxypyrazine 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-Ethynyl-5-methoxy-1,4-diazine(5-Methoxypyrazin-2-yl)acetylene

BLDPBD636480-250MGEA 149 EUR
BLDPBD636480-250MG BLDPBD636480-100MG BLDPBD636480-1G
2-Ethynyl-5-methoxypyrazine 97%
2-Ethynyl-5-methoxypyrazine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆N₂O
MW: 134,135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22199320
CAS Number: 1374115-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR