2-Bromo-2'-(trifluoromethyl)acetophenone 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone2-(Trifluoromethyl)phenacyl bromide

BLDPBD63706-250MGEA 30.5 EUR
BLDPBD63706-250MG BLDPBD63706-1G BLDPBD63706-10G BLDPBD63706-5G
2-Bromo-2'-(trifluoromethyl)acetophenone 98%
2-Bromo-2'-(trifluoromethyl)acetophenone

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆BrF₃O
MW: 267,04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094304
CAS Number: 54109-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR