2,2'-Dihydroxy-4-methoxybenzophenone 98%

Synonyms: Dioxybenzone,(2-Hydroxy-4-methoxyphenyl)(2-hydroxyphenyl)methanone

BD64469-100G BD64469-5G BD64469-25G
BLDPBD64469-100GEA 151 EUR
BLDPBD64469-100G BLDPBD64469-5G BLDPBD64469-25G
2,2'-Dihydroxy-4-methoxybenzophenone 98%
2,2'-Dihydroxy-4-methoxybenzophenone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₁₂O₄
MW: 244,246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002218
CAS Number: 131-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR