1,4-Benzodioxane-5-boronic acid 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: (2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)boronic acid

BLDPBD65597-1GEA 512 EUR
BLDPBD65597-1G BLDPBD65597-5G
1,4-Benzodioxane-5-boronic acid 95%
1,4-Benzodioxane-5-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₈H₉BO₄
MW: 179,966 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03095151
CAS Number: 499769-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR