5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%

Synonyms: (5-Chloro-2-(ethoxycarbonyl)phenyl)boronic acid5-Chloro-2-(ethoxycarbonyl)benzeneboronic acid

BLDPBD6572-250MGEA 71.4 EUR
BLDPBD6572-250MG BLDPBD6572-1G
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid 98%
5-Chloro-(2-ethoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₁₀BClO₄
MW: 228,44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363744
CAS Number: 871329-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR