Quinoxaline-2,3-dione 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2,3-DihydroxyquinoxalineQuinoxaline-2,3(1H,4H)-dione

BLDPBD65749-25GEA 32.6 EUR
BLDPBD65749-25G BLDPBD65749-100G BLDPBD65749-5G BLDPBD65749-10G
Quinoxaline-2,3-dione 98%
Quinoxaline-2,3-dione

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₆N₂O₂
MW: 162,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006723
CAS Number: 15804-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR