2',3'-Dimethoxyacetophenone 97%

BD65766-100MG BD65766-1G BD65766-250MG BD65766-5G
BLDPBD65766-100MGEA 14.7 EUR
BLDPBD65766-100MG BLDPBD65766-1G BLDPBD65766-250MG BLDPBD65766-5G
2',3'-Dimethoxyacetophenone 97%
2',3'-Dimethoxyacetophenone
Formula: C₁₀H₁₂O₃
MW: 180,2005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424412
CAS Number: 38480-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR