4-Bromo-3,5-dimethylbenzamide 97%

BD6580-1G BD6580-5G BD6580-25G
BLDPBD6580-1GEA 26.3 EUR
BLDPBD6580-1G BLDPBD6580-5G BLDPBD6580-25G
4-Bromo-3,5-dimethylbenzamide 97%
4-Bromo-3,5-dimethylbenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₀BrNO
MW: 228,0900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369913
CAS Number: 864825-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR