1-(2-Tetrahydrofuroyl)piperazine hydrobromide 97%

Synonyms: Piperazin-1-yl(tetrahydrofuran-2-yl)methanone hydrobromide

BLDPBD7056-5GEA 64.1 EUR
BLDPBD7056-5G BLDPBD7056-25G BLDPBD7056-1G
1-(2-Tetrahydrofuroyl)piperazine hydrobromide 97%
1-(2-Tetrahydrofuroyl)piperazine hydrobromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₇BrN₂O₂
MW: 265,1475 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701619
CAS Number: 63590-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR