2-Fluoro-1-methylpyridinium-p-toluenesulphonate 98%

Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate

BLDPBD74762-25GEA 128 EUR
BLDPBD74762-25G BLDPBD74762-1G BLDPBD74762-5G
2-Fluoro-1-methylpyridinium-p-toluenesulphonate 98%
2-Fluoro-1-methylpyridinium p-toluenesulphonate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₃H₁₄FNO₃S
MW: 283,32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011983
CAS Number: 58086-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR