2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%

Synonyms: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

BD74791-5G BD74791-10G BD74791-1G BD74791-25G BD74791-100G
BLDPBD74791-5GEA 39.9 EUR
BLDPBD74791-5G BLDPBD74791-10G BLDPBD74791-1G BLDPBD74791-25G BLDPBD74791-100G
2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-3-(trifluoromethyl)phenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅BF₄O₂
MW: 207,92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784336
CAS Number: 157834-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR