3-(4-Bromophenyl)propiononitrile 95%

Supplier: BLD PHARMATECH GMBH

Synonyms: 3-(4-Bromophenyl)propanenitrile

BLDPBD7520-10GEA 801 EUR
BLDPBD7520-10G BLDPBD7520-5G BLDPBD7520-100MG BLDPBD7520-1G BLDPBD7520-250MG
3-(4-Bromophenyl)propiononitrile 95%
3-(4-Bromophenyl)propiononitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₈BrN
MW: 210,0705 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01310795
CAS Number: 57775-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR