Fmoc-NH-PEG5-CH2COOH 96%

BD755734-5G BD755734-250MG BD755734-100MG BD755734-1G
BLDPBD755734-5GEA 1620 EUR
BLDPBD755734-5G BLDPBD755734-250MG BLDPBD755734-100MG BLDPBD755734-1G
Fmoc-NH-PEG5-CH2COOH 96%
Fmoc-NH-PEG5-CH2COOH
Formula: C₂₇H₃₅NO₉
MW: 517,568 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26142982
CAS Number: 635287-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR