6-Nitro-o-anisidine 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-Methoxy-6-nitrophenylamine2-Methoxy-6-nitroaniline

BLDPBD76451-5GEA 42 EUR
BLDPBD76451-5G BLDPBD76451-25G BLDPBD76451-250MG BLDPBD76451-500G BLDPBD76451-10G BLDPBD76451-100G BLDPBD76451-1G
6-Nitro-o-anisidine 98%
6-Nitro-o-anisidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₈N₂O₃
MW: 168,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01930197
CAS Number: 16554-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR