5-Bromo-2,4-dimethoxybenzaldehyde 98%

BLDPBD7676-250MGEA 17.9 EUR
BLDPBD7676-250MG BLDPBD7676-5G BLDPBD7676-1G
5-Bromo-2,4-dimethoxybenzaldehyde 98%
5-Bromo-2,4-dimethoxybenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₉BrO₃
MW: 245,0700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00010863
CAS Number: 130333-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR