2-Methylhippuric acid 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: 2-(2-Methylbenzamido)acetic acid

BLDPBD77266-5GEA 55.7 EUR
BLDPBD77266-5G BLDPBD77266-25G BLDPBD77266-1G
2-Methylhippuric acid 97%
2-Methylhippuric acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₁NO₃
MW: 193,1992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050991
CAS Number: 42013-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR