Isopropyldiphenylphosphine 98%

BLDPBD7728-250MGEA 24.2 EUR
BLDPBD7728-250MG BLDPBD7728-5G BLDPBD7728-1G
Isopropyldiphenylphosphine 98%
Isopropyldiphenylphosphine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₁₇P
MW: 228,27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00015028
CAS Number: 6372-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR