2-Undecylimidazole 96%
|
|
Synonyms:
2-Undecyl-1H-imidazole
BLDPBD77835-100GEA
53.6
EUR
BLDPBD77835-100G
BLDPBD77835-25G
BLDPBD77835-500G
BLDPBD77835-5G
BLDPBD77835-1000G
BLDPBD77835-0.025G
2-Undecylimidazole 96%
2-Undecylimidazole
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₄H₂₆N₂ MW: 222,3696 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038694 CAS Number: 16731-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...