Thiophene-2-carboxamide 98%

Synonyms: 2-Thiophenecarboxamide

BD78774-25G BD78774-5G BD78774-1G
BLDPBD78774-25GEA 128 EUR
BLDPBD78774-25G BLDPBD78774-5G BLDPBD78774-1G
Thiophene-2-carboxamide 98%
Thiophene-2-carboxamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₅NOS
MW: 127,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010425
CAS Number: 5813-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR