2-Chloro-6-(trifluoromethyl)phenylboronic acid 97%
|
|
BLDPBD8040-250MGEA
65.1
EUR
BLDPBD8040-250MG
BLDPBD8040-1G
BLDPBD8040-100MG
2-Chloro-6-(trifluoromethyl)phenylboronic acid 97%
2-Chloro-6-(trifluoromethyl)phenylboronic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₇H₅BClF₃O₂ MW: 224,3726 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09037968 CAS Number: 851756-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...