3-[(Methylsulphonyl)amino]benzoic acid 98%

Synonyms: 3-(Methylsulphonamido)benzoic acid,N-(3-Carboxyphenyl)methanesulphonamide

BD80722-250MG BD80722-5G BD80722-10G BD80722-1G
BLDPBD80722-250MGEA 34.7 EUR
BLDPBD80722-250MG BLDPBD80722-5G BLDPBD80722-10G BLDPBD80722-1G
3-[(Methylsulphonyl)amino]benzoic acid 98%
3-[(Methylsulphonyl)amino]benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₉NO₄S
MW: 215,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00596197
CAS Number: 28547-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR