3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 4,4,5,5-tetramethyl-2-(3-((tetrahydro-2H-pyran-2-yl)oxy)phenyl)-1,3,2-dioxaborolane

BLDPBD80974-5GEA 178 EUR
BLDPBD80974-5G BLDPBD80974-1G BLDPBD80974-250MG
3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester 98%
3-(2-Tetrahydro-2H-pyran-2-yloxy)benzeneboronic acid, pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₇H₂₅BO₄
MW: 304,189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659834
CAS Number: 850568-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR